Assay Details
Target Gene Details
Entrez Gene ID: | 22061 |
Gene Name: | transformation related protein 63 |
Gene Aliases: |
AI462811, Ket, P51/P63, P63, P73l, Tp63, Trp53rp1 |
Location: |
Chr.16:25683763-25892103 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Trp63 | NM_001127259.1 | 2 | 653 | NP_001120731.1 |
NM_001127260.1 | 2 | 653 | NP_001120732.1 | |
NM_001127261.1 | 2 | 653 | NP_001120733.1 | |
AF075434.1 | 2 | 128 | AAC62639.1 | |
AF075435.1 | 2 | 128 | AAC62640.1 | |
AF075436.1 | 2 | 128 | AAC62641.1 | |
AK138305.1 | 2 | 657 |
More Information
Set Membership: |
Intragenic Exonic |