Assay Details
Target Gene Details
Entrez Gene ID: | 208177 |
Gene Name: | pleckstrin homology-like domain, family B, member 2 |
Gene Aliases: |
AV253284, C820004H04Rik, LL5b, LL5beta |
Location: |
Chr.16:45746230-45953701 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Phldb2 | NM_001252442.1 | 5 | 2101 | NP_001239371.1 |
NM_153412.4 | 5 | 2101 | NP_700461.2 | |
AF506821.1 | 5 | 2102 | AAM33635.1 | |
AK035655.1 | BAC29141.1 | |||
AK049595.1 | 5 | 1955 | BAC33833.1 | |
AK050493.1 | 6 | 2268 | ||
AK165251.1 | 5 | 2063 | BAE38105.1 | |
BC050915.1 | 5 | 1957 | AAH50915.1 | |
BC060683.1 | 5 | 1957 | AAH60683.1 |