Assay Details
Target Gene Details
Entrez Gene ID: | 239719 |
Gene Name: | MKL/myocardin-like 2 |
Gene Aliases: |
ENSMUSG00000075401, Gt4-1, MRTF-B, Mrtfb, mKIAA1243 |
Location: |
Chr.16:13256017-13417529 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mkl2 | NM_001122667.2 | NP_001116139.1 | ||
NM_153588.3 | NP_705816.2 | |||
AF532598.1 | AAN33042.1 | |||
AK220377.1 | BAD90435.1 | |||
BC131644.1 | ||||
BC131645.1 |
More Information
Set Membership: |
Intragenic Non-exonic |