Assay Details
Target Gene Details
Entrez Gene ID: | 210126 |
Gene Name: | LIM domain containing preferred translocation partner in lipoma |
Gene Aliases: |
9430020K16Rik, AA959454, AU024130, B130055L10Rik, C79715, D630048H16 |
Location: |
Chr.16:24393350-24992578 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lpp | NM_001145952.1 | NP_001139424.1 | ||
NM_001145954.1 | NP_001139426.1 | |||
NM_178665.5 | NP_848780.3 | |||
AK029335.1 | BAC26401.1 | |||
AK029567.1 | BAC26516.1 | |||
AK040643.1 | BAC30654.1 | |||
AK045288.1 | BAC32298.1 | |||
AK051345.1 | BAC34608.1 | |||
AK054550.1 | BAC35821.1 | |||
AK076989.1 | BAC36552.1 | |||
AK085626.1 | ||||
AK086307.1 | ||||
AK168431.1 | ||||
AK169780.1 | BAE41362.1 | |||
BC085321.1 | AAH85321.1 |
More Information
Set Membership: |
Intragenic Non-exonic |