Assay Details
Target Gene Details
Entrez Gene ID: | 12394 |
Gene Name: | runt related transcription factor 1 |
Gene Aliases: |
AML1, CBF-alpha-2, Cbfa2, Pebp2a2, Pebpa2b |
Location: |
Chr.16:92601466-92826074 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Runx1 | NM_001111021.2 | NP_001104491.1 | ||
NM_001111022.2 | NP_001104492.1 | |||
AF345649.1 | AAK29784.1 | |||
AK040240.1 | ||||
AK137493.1 | ||||
AK145098.1 | BAE26233.1 | |||
AK165307.1 | ||||
BC069929.1 | AAH69929.1 | |||
BF135151.1 | ||||
W29200.1 |
More Information
Set Membership: |
Intragenic Non-exonic |