Assay Details
Target Gene Details
Entrez Gene ID: | 18203 |
Gene Name: | N-terminal Asn amidase |
Gene Aliases: |
- |
Location: |
Chr.16:13818834-13835450 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ntan1 | NM_010946.3 | NP_035076.1 | ||
AK041112.1 | BAC30825.1 | |||
AK207258.1 | ||||
AK214245.1 | ||||
BC030172.1 | AAH30172.1 | |||
BY254590.1 | ||||
U57692.1 | AAB66490.1 |
More Information
Set Membership: |
Intragenic Non-exonic |