Assay Details
Target Gene Details
Entrez Gene ID: | 21762 |
Gene Name: | proteasome (prosome, macropain) 26S subunit, non-ATPase, 2 |
Gene Aliases: |
9430095H01Rik, AA407121, TEG-190, Tex190 |
Location: |
Chr.16:20651652-20663414 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 19 - Intron 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Psmd2 | NM_134101.2 | NP_598862.1 | ||
NR_027485.1 | ||||
AK020524.1 | BAB32124.1 | |||
AK146482.1 | ||||
AK150374.1 | BAE29506.1 | |||
AK151588.1 | BAE30528.1 | |||
AK152007.1 | BAE30871.1 | |||
AK152970.1 | ||||
AK159098.1 | BAE34814.1 | |||
AK159636.1 | BAE35250.1 | |||
AK159756.1 | BAE35347.1 | |||
AK159815.1 | BAE35395.1 | |||
AK166816.1 | BAE39042.1 | |||
AK167326.1 | BAE39428.1 | |||
AK167774.1 | BAE39806.1 | |||
AK167786.1 | ||||
AK167909.1 | BAE39916.1 | |||
AK167995.1 | BAE39985.1 | |||
AK168131.1 | BAE40100.1 | |||
AK168398.1 | ||||
AK168543.1 | ||||
AK169867.1 | ||||
BC005592.1 | AAH05592.1 | |||
BC021508.1 | AAH21508.1 | |||
BC023049.1 | AAH23049.1 | |||
BC024830.1 | AAH24830.1 | |||
BC027388.1 | AAH27388.1 | |||
BC027767.1 | AAH27767.1 | |||
BC027822.1 | AAH27822.1 | |||
BC028851.1 | AAH28851.1 | |||
BC031128.1 | AAH31128.1 |
More Information
Set Membership: |
Intragenic Non-exonic |