Assay Details
Target Gene Details
Entrez Gene ID: | 72599 |
Gene Name: | protein disulfide isomerase associated 5 |
Gene Aliases: |
2700053F16Rik, AU015525, Pdir |
Location: |
Chr.16:35397308-35490898 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pdia5 | NM_028295.1 | 17 | 1695 | NP_082571.1 |
AK012415.1 | 5 | 486 | BAB28225.1 | |
BC009151.1 | 17 | 1695 | AAH09151.1 |
More Information
Set Membership: |
Intragenic Exonic |