Assay Details
Target Gene Details
Entrez Gene ID: | 207781 |
Gene Name: | C2 calcium-dependent domain containing 2 |
Gene Aliases: |
5730563M15, 5830404H04Rik, AI314146, AI449533, BF168443, ORF25, mKIAA4045 |
Location: |
Chr.16:97855209-97922651 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C2cd2 | NM_174847.2 | NP_777272.2 | ||
AK017896.1 | ||||
AK077731.1 | ||||
AK080956.1 | ||||
AK086520.1 | ||||
AK143875.1 | BAE25577.1 | |||
AK169902.1 | BAE41447.1 | |||
AK220454.1 | BAD90483.1 | |||
BC043688.1 | AAH43688.1 |
Target Gene Details
Entrez Gene ID: | 102465581 |
Gene Name: | microRNA 6964 |
Gene Aliases: |
mmu-mir-6964 |
Location: |
Chr.16:97877233-97877291 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mir6964 | NR_105929.1 |