Assay Details
Target Gene Details
Entrez Gene ID: | 22762 |
Gene Name: | zinc finger protein, multitype 2 |
Gene Aliases: |
B330005D23Rik, FOG-2, FOG2 |
Location: |
Chr.15:40655042-41104592 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zfpm2 | NM_011766.5 | NP_035896.1 | ||
AF107306.1 | AAD12182.1 | |||
AF118845.1 | AAD17251.1 | |||
AF125166.1 | AAD33250.1 | |||
AK046521.1 | ||||
BC059241.1 | AAH59241.1 | |||
EF514219.1 |
More Information
Set Membership: |
Intragenic Non-exonic |