Assay Details
Target Gene Details
Entrez Gene ID: | 22762 |
Gene Name: | zinc finger protein, multitype 2 |
Gene Aliases: |
B330005D23Rik, FOG-2, FOG2 |
Location: |
Chr.15:40655042-41104592 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zfpm2 | NM_011766.5 | 3 | 757 | NP_035896.1 |
AF107306.1 | 3 | 278 | AAD12182.1 | |
AF118845.1 | 3 | 527 | AAD17251.1 | |
AF125166.1 | 3 | 758 | AAD33250.1 | |
AK033131.1 | 3 | 449 | BAC28166.1 | |
AK046521.1 | 3 | 765 | ||
BC059241.1 | 3 | 657 | AAH59241.1 | |
EF514219.1 | 3 | 1580 |
More Information
Set Membership: |
Intragenic Exonic |