Assay Details
Target Gene Details
Entrez Gene ID: | 223669 |
Gene Name: | zinc finger protein 7 |
Gene Aliases: |
KRAB20, KRAB7, Krox-2, Zfp-7, Zfp65, Zfp80, Zfp86-rs1, mszf73-2 |
Location: |
Chr.15:76879217-76892395 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zfp7 | NM_145916.2 | NP_666021.2 | ||
AK143414.1 | ||||
AK144887.1 | BAE26117.1 | |||
AK168948.1 | BAE40754.1 | |||
BC011501.1 | AAH11501.1 | |||
BU613201.1 | ||||
U29499.1 | AAC52294.1 |