Assay Details
Target Gene Details
Entrez Gene ID: | 14390 |
Gene Name: | GA repeat binding protein, alpha |
Gene Aliases: |
GABPalpha |
Location: |
Chr.16:84834925-84863783 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gabpa | NM_008065.2 | 8 | 1357 | NP_032091.2 |
AK139916.1 | 8 | 1037 | BAE24181.1 | |
AK145446.1 | 8 | 1037 | BAE26442.1 | |
AK145838.1 | 8 | 1102 | BAE26687.1 | |
AK148087.1 | 8 | 1128 | BAE28337.1 | |
AK150455.1 | 8 | 1037 | BAE29575.1 | |
AY282796.1 | 1 | 109 | AAQ19481.1 | |
AY282797.1 | 1 | 109 | AAQ19482.1 | |
AY282798.1 | 1 | 109 | AAQ19483.1 | |
AY282799.1 | 1 | 109 | AAQ19484.1 | |
BC013562.1 | 6 | 752 | AAH13562.1 | |
BC052448.1 | 8 | 1357 | AAH52448.1 | |
FN545817.1 | 3 | 194 | ||
M74515.1 | 8 | 1364 | AAA53030.1 |
More Information
Set Membership: |
Intragenic Exonic |