Assay Details
Target Gene Details
Entrez Gene ID: | 72555 |
Gene Name: | shisa family member 9 |
Gene Aliases: |
2700045P11Rik, Ckamp44 |
Location: |
Chr.16:11984113-12270904 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Shisa9 | NM_001174086.1 | NP_001167557.1 | ||
NM_028277.2 | NP_082553.2 | |||
AK049860.1 | ||||
GU479981.1 | ||||
GU479982.1 |