Assay Details
Target Gene Details
Entrez Gene ID: | 71753 |
Gene Name: | transmembrane serine protease 6 |
Gene Aliases: |
1300008A22Rik |
Location: |
Chr.15:78439667-78468648 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tmprss6 | NM_027902.2 | NP_082178.2 | ||
AK004939.2 | BAB23684.2 | |||
AY240929.1 | AAP69827.1 | |||
BC029645.1 | AAH29645.2 | |||
BC057674.1 | AAH57674.1 | |||
EU190436.1 | ||||
EU190437.1 |
More Information
Set Membership: |
Intragenic Non-exonic |