Assay Details
Target Gene Details
Entrez Gene ID: | 268860 |
Gene Name: | 4-aminobutyrate aminotransferase |
Gene Aliases: |
9630038C02Rik, AI255750, ENSMUSG00000051226, Gabaat, Gabat, Gm9851, I54, Laibat, X61497 |
Location: |
Chr.16:8513429-8621568 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Abat | NM_001170978.1 | NP_001164449.1 | ||
NM_172961.3 | NP_766549.2 | |||
AK036128.1 | BAC29312.1 | |||
AK132282.1 | BAE21080.1 | |||
AK160802.1 | BAE36023.1 | |||
AK165348.1 | ||||
BC058079.1 | AAH58079.1 | |||
BC058521.1 | AAH58521.1 | |||
CX237932.1 |
More Information
Set Membership: |
Intragenic Non-exonic |