Assay Details
Target Gene Details
Entrez Gene ID: | 74374 |
Gene Name: | C-type lectin domain family 16, member A |
Gene Aliases: |
4932416N17Rik, curt, mKIAA0350 |
Location: |
Chr.16:10545339-10744878 on Build GRCm38 |
Assay Gene Location: | Within Exon 23 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Clec16a | NM_001204229.1 | 22 | 2846 | NP_001191158.1 |
NM_177562.5 | 23 | 2852 | NP_808230.2 | |
AK016529.1 | 13 | 1541 | BAC25487.1 | |
AK122255.1 | 23 | 2804 | BAC65537.1 | |
AK156927.1 | 22 | 2777 | BAE33899.1 | |
AK171134.1 | 21 | 2704 | ||
BC138907.1 | 23 | 2755 | ||
BC138909.1 | 23 | 2755 |