Assay Details
Target Gene Details
Entrez Gene ID: | 223693 |
Gene Name: | transmembrane protein 184b |
Gene Aliases: |
2610507A11, 4732495E13Rik |
Location: |
Chr.15:79360684-79403303 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tmem184b | NM_001253817.1 | 6 | 702 | NP_001240746.1 |
NM_001253819.1 | 6 | 723 | NP_001240748.1 | |
NM_001253820.1 | 4 | 268 | NP_001240749.1 | |
NM_172608.1 | 6 | 748 | NP_766196.1 | |
AK028196.1 | 6 | 706 | BAC25805.1 | |
AK029129.1 | 6 | 726 | BAC26312.1 | |
AK048138.1 | 6 | 702 | BAC33254.1 | |
AK149366.1 | 6 | 729 | BAE28837.1 | |
AK158789.1 | 6 | 749 | BAE34666.1 | |
AK170086.1 | 6 | 726 | BAE41556.1 | |
AK202130.1 | 2 | 103 | ||
BC046959.1 | 6 | 702 | AAH46959.1 | |
CX239857.1 | 4 | 268 |
More Information
Set Membership: |
Intragenic Exonic |