Assay Details
Target Gene Details
Entrez Gene ID: | 12469 |
Gene Name: | chaperonin containing Tcp1, subunit 8 (theta) |
Gene Aliases: |
AI132397, Cctq, Tcpq |
Location: |
Chr.16:87483325-87495869 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cct8 | NM_009840.3 | NP_033970.3 | ||
AK014005.1 | BAB29110.1 | |||
AK075711.1 | ||||
AK075880.1 | BAC36025.1 | |||
AK145661.1 | BAE26573.1 | |||
AK145754.1 | BAE26629.1 | |||
AK145918.1 | BAE26749.1 | |||
AK147038.1 | BAE27627.1 | |||
AK159969.1 | ||||
AK168465.1 | ||||
AK172867.1 | BAD32145.1 | |||
BC009007.1 | AAH09007.1 | |||
Z37164.1 | CAA85521.1 |
More Information
Set Membership: |
Intragenic Non-exonic |