Assay Details
Target Gene Details
Entrez Gene ID: | 239706 |
Gene Name: | methyltransferase like 22 |
Gene Aliases: |
- |
Location: |
Chr.16:8470753-8490205 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mettl22 | NM_146247.1 | 2 | 121 | NP_666359.1 |
AK089212.1 | 2 | 136 | ||
AK136614.1 | 2 | 147 | BAE23071.1 | |
BC024814.1 | 2 | 121 | AAH24814.1 |
More Information
Set Membership: |
Intragenic Exonic |