Assay Details
Target Gene Details
Entrez Gene ID: | 75329 |
Gene Name: | activating transcription factor 7 interacting protein 2 |
Gene Aliases: |
4930558K11Rik, BC018510, Get-1, PSM2 |
Location: |
Chr.16:10192640-10249669 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Atf7ip2 | NM_029253.1 | NP_083529.1 | ||
NM_153123.2 | NP_694763.2 | |||
AK016180.1 | BAB30138.1 | |||
AK077229.1 | BAC36698.1 | |||
AK145634.1 | BAE26553.1 | |||
AY903215.1 | AAX99226.1 |
More Information
Set Membership: |
Intragenic Non-exonic |