Assay Details
Target Gene Details
Entrez Gene ID: | 20286 |
Gene Name: | zinc finger CCCH type containing 7B |
Gene Aliases: |
Gm627, Scrg3, Zc2h7b, mKIAA1031 |
Location: |
Chr.15:81744848-81796269 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zc3h7b | NM_001081016.1 | NP_001074485.1 | ||
AK129268.1 | BAC98078.2 | |||
AK147484.1 | ||||
AK147978.1 | BAE28263.1 | |||
AK165492.1 | ||||
AK171949.1 | BAE42742.1 |
More Information
Set Membership: |
Intragenic Non-exonic |