Assay Details
Target Gene Details
Entrez Gene ID: | 70508 |
Gene Name: | bobby sox homolog (Drosophila) |
Gene Aliases: |
5530401J07Rik, 5730403O13Rik |
Location: |
Chr.16:50191844-50432461 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bbx | NM_027444.3 | 13 | 2393 | NP_081720.2 |
AF454943.1 | 13 | 2386 | AAL58872.1 | |
AF454944.1 | 11 | 2258 | AAL58873.1 | |
AK029532.1 | BAC26500.2 | |||
AK029747.1 | 14 | 2507 | BAC26596.1 | |
AK041632.1 | BAC31013.1 | |||
AK049516.1 | 13 | 2379 | BAC33788.1 | |
AK167724.1 | 13 | 2524 | ||
BC066821.1 | 11 | 2020 | AAH66821.1 |
More Information
Set Membership: |
Intragenic Exonic |