Assay Details
Target Gene Details
Entrez Gene ID: | 320214 |
Gene Name: | MYCBP-associated, testis expressed 1 |
Gene Aliases: |
4932425I24Rik, Aat1, Spata26 |
Location: |
Chr.16:38297754-38342253 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Maats1 | NM_001081025.1 | 1 | 152 | NP_001074494.1 |
AF503943.1 | 1 | 435 | AAM27269.1 | |
AK045121.1 | 1 | 104 | BAC32231.1 | |
AK077038.1 | 1 | 138 |