Assay Details
Target Gene Details
Entrez Gene ID: | 224045 |
Gene Name: | eukaryotic translation initiation factor 2B, subunit 5 epsilon |
Gene Aliases: |
C81315 |
Location: |
Chr.16:20498808-20509325 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 10 - Intron 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Eif2b5 | NM_172265.2 | NP_758469.1 | ||
AK145801.1 | ||||
AK160985.1 | BAE36132.1 | |||
BC038620.1 | AAH38620.1 | |||
BC085255.1 | AAH85255.1 |
More Information
Set Membership: |
Intragenic Non-exonic |