Assay Details
Target Gene Details
Entrez Gene ID: | 12667 |
Gene Name: | chordin |
Gene Aliases: |
Chd |
Location: |
Chr.16:20731417-20742384 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 22 - Intron 22 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Chrd | NM_001278041.1 | NP_001264970.1 | ||
NM_009893.2 | NP_034023.1 | |||
AF069501.1 | AAC68867.1 | |||
AF096276.1 | AAD19895.1 | |||
AK077460.1 | ||||
AK149445.1 | BAE28881.1 | |||
AK220310.1 | 5 | 3610 | BAD90387.1 | |
BC145696.1 |