Assay Details
Target Gene Details
Entrez Gene ID: | 56297 |
Gene Name: | ADP-ribosylation factor-like 6 |
Gene Aliases: |
1110018H24Rik, 2210411E14Rik, BBS3 |
Location: |
Chr.16:59613321-59639405 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Arl6 | NM_019665.3 | NP_062639.3 | ||
AB232697.1 | BAE79900.1 | |||
AF031903.1 | AAC62194.1 | |||
AK003778.1 | BAB22990.1 | |||
AK152980.1 | BAE31631.1 | |||
AK158832.1 | BAE34686.1 | |||
AK160666.1 | BAE35949.1 | |||
BC018497.1 | AAH18497.1 |
More Information
Set Membership: |
Intragenic Non-exonic |