Assay Details
Target Gene Details
Entrez Gene ID: | 11815 |
Gene Name: | apolipoprotein D |
Gene Aliases: |
- |
Location: |
Chr.16:31296192-31314808 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Apod | NM_001301353.1 | 3 | 407 | NP_001288282.1 |
NM_001301354.1 | 2 | 341 | NP_001288283.1 | |
NM_007470.4 | 3 | 411 | NP_031496.2 | |
AK135046.1 | 3 | 199 | BAE22397.1 | |
AK157917.1 | 3 | 200 | BAE34262.1 | |
AK157925.1 | 3 | 199 | ||
AK158118.1 | 3 | 196 | BAE34364.1 | |
AK158405.1 | 3 | 202 | BAE34491.1 | |
AK160729.1 | 2 | 130 | BAE35974.1 | |
AK162392.1 | 3 | 199 | BAE36889.1 | |
BC145907.1 | 1 | 89 | ||
BC145909.1 | 1 | 89 | ||
X82648.1 | 2 | 145 | CAA57974.1 |
More Information
Set Membership: |
Intragenic Exonic |