Assay Details
Target Gene Details
Entrez Gene ID: | 11815 |
Gene Name: | apolipoprotein D |
Gene Aliases: |
- |
Location: |
Chr.16:31296192-31314808 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Apod | NM_001301353.1 | NP_001288282.1 | ||
NM_001301354.1 | NP_001288283.1 | |||
NM_007470.4 | NP_031496.2 | |||
AK135046.1 | BAE22397.1 | |||
AK157917.1 | BAE34262.1 | |||
AK157925.1 | ||||
AK158118.1 | BAE34364.1 | |||
AK158405.1 | BAE34491.1 | |||
AK160729.1 | BAE35974.1 | |||
AK162392.1 | BAE36889.1 |
More Information
Set Membership: |
Intragenic Non-exonic |