Assay Details
Target Gene Details
Entrez Gene ID: | 208650 |
Gene Name: | Casitas B-lineage lymphoma b |
Gene Aliases: |
AI429560, AI851073, Cbl-b |
Location: |
Chr.16:52031230-52208047 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cblb | NM_001033238.1 | NP_001028410.1 | ||
AK147367.1 | ||||
AK156295.1 | ||||
AK161486.1 | BAE36418.1 | |||
AK170640.1 | ||||
AK171350.1 | ||||
BC150934.1 | ||||
BC150938.1 |
More Information
Set Membership: |
Intragenic Non-exonic |