Assay Details
Target Gene Details
Entrez Gene ID: | 11964 |
Gene Name: | ATPase, H+ transporting, lysosomal V1 subunit A |
Gene Aliases: |
AI647066, Atp6a1, Atp6a2, Atp6v1a1, VA68, VPP2 |
Location: |
Chr.16:44085402-44139700 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Atp6v1a | NM_007508.5 | NP_031534.2 | ||
AK140873.1 | BAE24506.1 | |||
AK149833.1 | BAE29112.1 | |||
AK150742.1 | ||||
AK153403.1 | BAE31963.1 | |||
AK154869.1 | BAE32890.1 | |||
AK160792.1 | BAE36015.1 | |||
AK166857.1 | BAE39074.1 | |||
AK170721.1 | BAE41978.1 | |||
AK211144.1 | ||||
BC038392.1 | AAH38392.1 |
More Information
Set Membership: |
Intragenic Non-exonic |