Assay Details
Target Gene Details
Entrez Gene ID: | 66194 |
Gene Name: | pyrroline-5-carboxylate reductase-like |
Gene Aliases: |
1110058B13Rik, 2700073G24Rik |
Location: |
Chr.15:75916463-75921560 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pycrl | NM_025412.2 | 4 | 497 | NP_079688.2 |
AK002912.1 | 4 | 428 | BAB22451.1 | |
AK004291.1 | 4 | 501 | BAB23252.1 | |
AK013214.1 | 4 | 423 | ||
BC026536.1 | 4 | 418 | AAH26536.1 | |
CX212991.1 |
More Information
Set Membership: |
Intragenic Exonic |