Assay Details
Target Gene Details
Entrez Gene ID: | 223754 |
Gene Name: | TBC1 domain family, member 22a |
Gene Aliases: |
BC023106, D15Ertd781e |
Location: |
Chr.15:86214421-86498503 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tbc1d22a | NM_145476.2 | 9 | 1183 | NP_663451.2 |
AK039627.1 | 9 | 1196 | BAC30403.1 | |
AK146872.1 | 9 | 1184 | BAE27495.1 | |
AK154226.1 | 9 | 1205 | BAE32447.1 | |
AK154602.1 | 9 | 1226 | BAE32705.1 | |
AK155459.1 | 9 | 1197 | BAE33274.1 | |
AK155770.1 | 9 | 1226 | BAE33429.1 | |
BC023106.1 | 6 | 578 | AAH23106.1 | |
BC066009.1 | 9 | 1183 | AAH66009.1 |
More Information
Set Membership: |
Intragenic Exonic |