Assay Details
Target Gene Details
Entrez Gene ID: | 19011 |
Gene Name: | endonuclease, polyU-specific |
Gene Aliases: |
Pp11r, Tcl-30 |
Location: |
Chr.15:97711015-97731405 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Endou | NM_001168693.1 | NP_001162164.1 | ||
NM_008902.3 | NP_032928.1 | |||
AK132616.1 | BAE21263.1 | |||
AK169457.1 | ||||
M95545.1 | AAA40405.1 |