Assay Details
Target Gene Details
Entrez Gene ID: | 69572 |
Gene Name: | major facilitator superfamily domain containing 3 |
Gene Aliases: |
2310010G13Rik |
Location: |
Chr.15:76701542-76704239 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mfsd3 | NM_027122.3 | NP_081398.2 | ||
AK009269.1 | BAB26184.1 | |||
AK089539.1 | BAC40919.1 | |||
AK217168.1 | ||||
BC019171.1 | AAH19171.1 | |||
BC085305.1 | AAH85305.1 |
More Information
Set Membership: |
Intragenic Non-exonic |