Assay Details
Target Gene Details
Entrez Gene ID: | 105837 |
Gene Name: | Mdm2, transformed 3T3 cell double minute p53 binding protein |
Gene Aliases: |
AI429604, MDM2BP |
Location: |
Chr.15:55557408-55626423 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mtbp | NM_001168250.1 | 4 | 562 | NP_001161722.1 |
NM_134092.3 | 4 | 562 | NP_598853.3 | |
AJ278508.1 | 4 | 483 | CAC07970.1 | |
AK080036.1 | 4 | 376 | BAC37810.1 | |
AK088549.1 | 4 | 376 | BAC40418.1 | |
AK133855.1 | 4 | 410 | BAE21889.1 | |
BB615361.1 | 4 | 538 | ||
BC052197.1 | 4 | 562 | AAH52197.1 |