Assay Details
Target Gene Details
Entrez Gene ID: | 239510 |
Gene Name: | PHD finger protein 20-like 1 |
Gene Aliases: |
CGI-72, E130113K22Rik |
Location: |
Chr.15:66577517-66647043 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Phf20l1 | NM_001081409.1 | 12 | 1827 | NP_001074878.1 |
AK032638.1 | 12 | 1842 | BAC27964.1 | |
AK040888.1 | 3 | 558 | BAC30733.1 | |
AK041825.1 | 2 | 922 | ||
AK053606.1 | 1 | 63 | BAC35444.1 | |
AK080555.1 | 2 | 1459 | BAC37943.1 | |
AK187831.1 | 1 | 124 | ||
BC052212.1 | 11 | 1753 | AAH52212.1 | |
BC061492.1 | 11 | 1693 | AAH61492.1 | |
BC082777.1 | 12 | 1822 | AAH82777.1 | |
BC145689.1 | 11 | 1558 |