Assay Details
Target Gene Details
Entrez Gene ID: | 105835 |
Gene Name: | small G protein signaling modulator 3 |
Gene Aliases: |
1810012I01Rik, AI428557, BB175482, Cip85, R75178, RUSC3, Rutbc3 |
Location: |
Chr.15:80977765-81012290 on Build GRCm38 |
Assay Gene Location: | Within Intron 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sgsm3 | NM_134091.2 | NP_598852.2 | ||
AK007455.1 | ||||
AK142314.1 | ||||
AK170806.1 | BAE42041.1 | |||
AK183562.1 | ||||
AY382616.1 | AAR89983.1 | |||
BC018197.1 | AAH18197.1 | |||
BC024122.1 | AAH24122.1 | |||
BC025561.1 | AAH25561.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |