Assay Details
Target Gene Details
Entrez Gene ID: | 56612 |
Gene Name: | prefoldin 5 |
Gene Aliases: |
1190001O17Rik, 1700010A06Rik, D15Ertd697e, Eig1, MM-1 |
Location: |
Chr.15:102326116-102331490 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pfdn5 | NM_027044.3 | NP_081320.2 | ||
AF108357.1 | AAD28373.1 | |||
AK004438.1 | ||||
AK005682.1 | BAB24185.2 | |||
AK028206.1 | BAC25814.1 | |||
AK145883.1 | BAE26723.1 | |||
BC026920.1 | AAH26920.1 | |||
CN835522.1 |
More Information
Set Membership: |
Intragenic Non-exonic |