Assay Details
Target Gene Details
Entrez Gene ID: | 320506 |
Gene Name: | LMBR1 domain containing 2 |
Gene Aliases: |
9930036E21Rik |
Location: |
Chr.15:9140533-9202483 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lmbrd2 | NM_177178.3 | NP_796152.2 | ||
AK036527.1 | ||||
AK051777.1 | BAC34764.1 | |||
AK079450.1 | BAC37649.1 | |||
AK153728.1 | BAE32158.1 | |||
BC095971.1 | AAH95971.1 | |||
BC120727.1 | ||||
BC125421.1 |