Assay Details
Target Gene Details
Entrez Gene ID: | 320506 |
Gene Name: | LMBR1 domain containing 2 |
Gene Aliases: |
9930036E21Rik |
Location: |
Chr.15:9140533-9202483 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lmbrd2 | NM_177178.3 | 17 | 2187 | NP_796152.2 |
AK036527.1 | 17 | 2209 | ||
AK051777.1 | 11 | 1194 | BAC34764.1 | |
AK079450.1 | 17 | 2187 | BAC37649.1 | |
AK153728.1 | 17 | 2138 | BAE32158.1 | |
BC095971.1 | 17 | 1969 | AAH95971.1 | |
BC120727.1 | 17 | 2159 | ||
BC125421.1 | 17 | 2159 |