Assay Details
Target Gene Details
Entrez Gene ID: | 53357 |
Gene Name: | phospholipase A2, group VI |
Gene Aliases: |
BB112799, PNPLA9, iPLA(2)beta, iPLA2, iPLA2beta |
Location: |
Chr.15:79286228-79328530 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pla2g6 | NM_001199023.1 | 14 | 2739 | NP_001185952.1 |
NM_001199024.1 | 13 | 2574 | NP_001185953.1 | |
NM_001199025.1 | 14 | 2079 | NP_001185954.1 | |
NM_016915.4 | 13 | 2120 | NP_058611.1 | |
AF259401.1 | 13 | 2019 | AAF72651.1 | |
AK144495.1 | 14 | 1953 | BAE25917.1 | |
AK153333.1 | 13 | 2380 | BAE31912.1 | |
AK204420.1 | 1 | 56 | ||
BC003487.1 | 13 | 1913 | AAH03487.1 | |
BC052845.1 | 13 | 1945 | AAH52845.1 | |
BC057209.1 | 14 | 2332 | AAH57209.1 | |
U88624.2 | 12 | 1796 | AAB48511.2 |
More Information
Set Membership: |
Intragenic Exonic |