Assay Details
Target Gene Details
Entrez Gene ID: | 26942 |
Gene Name: | sperm associated antigen 1 |
Gene Aliases: |
tpis |
Location: |
Chr.15:36175483-36235621 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 14 - Intron 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Spag1 | NM_012031.3 | NP_036161.2 | ||
AF181252.1 | AAF06160.1 | |||
AF181253.1 | AAF06161.1 | |||
AK032601.1 | BAC27944.1 | |||
BC046313.1 | AAH46313.1 |