Assay Details
Target Gene Details
Entrez Gene ID: | 68877 |
Gene Name: | MAF1 homolog (S. cerevisiae) |
Gene Aliases: |
1110068E11Rik, AU042856 |
Location: |
Chr.15:76351294-76354378 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Maf1 | NM_001164607.1 | NP_001158079.1 | ||
NM_001164608.1 | NP_001158080.1 | |||
NM_026859.3 | NP_081135.3 | |||
AK004406.1 | BAB23294.1 | |||
AK042913.1 | BAC31403.1 | |||
AK154047.1 | BAE32337.1 | |||
AK189875.1 | ||||
AK196909.1 | ||||
BC016260.1 | AAH16260.1 |
More Information
Set Membership: |
Intragenic Non-exonic |