Assay Details
Target Gene Details
Entrez Gene ID: | 20512 |
Gene Name: | solute carrier family 1 (glial high affinity glutamate transporter), member 3 |
Gene Aliases: |
AI504299, B430115D02Rik, Eaat1, GLAST, GLAST-1, GLU-T, GluT-1, Gmt1, MGluT1 |
Location: |
Chr.15:8634124-8710807 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc1a3 | NM_148938.3 | NP_683740.1 | ||
AF330257.1 | AAK01708.1 | |||
AK031831.1 | ||||
AK040798.1 | BAC30705.1 | |||
AK045716.1 | ||||
AK045949.1 | BAC32544.1 | |||
AK049090.1 | BAC33538.1 | |||
AK080879.1 | ||||
AK082736.1 | BAC38592.1 | |||
AK084968.1 | BAC39324.1 | |||
AK158777.1 | BAE34658.1 | |||
BC058711.1 | AAH58711.1 | |||
BC066154.1 | AAH66154.1 |
More Information
Set Membership: |
Intragenic Non-exonic |