Assay Details
Target Gene Details
Entrez Gene ID: | 12984 |
Gene Name: | colony stimulating factor 2 receptor, beta 2, low-affinity (granulocyte-macrophage) |
Gene Aliases: |
AIC2A, BetaIl3, Bil3, Csfgmrb, Il3r, Il3rb, Il3rb2 |
Location: |
Chr.15:78282507-78305721 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Csf2rb2 | NM_001287389.1 | NP_001274318.1 | ||
NM_007781.3 | NP_031807.1 | |||
AK154650.1 | BAE32741.1 | |||
AK155216.1 | BAE33127.1 | |||
AK155511.1 | BAE33301.1 | |||
AK155626.1 | BAE33352.1 | |||
AK155675.1 | BAE33383.1 | |||
AK170028.1 | BAE41518.1 | |||
M29855.1 | AAA39295.1 |
More Information
Set Membership: |
Intragenic Non-exonic |