Assay Details
Target Gene Details
Entrez Gene ID: | 70363 |
Gene Name: | family with sequence similarity 135, member B |
Gene Aliases: |
1700010C24Rik, A830008O07Rik |
Location: |
Chr.15:71438951-71728436 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fam135b | NM_177819.3 | 18 | 4026 | NP_808487.2 |
AK005816.1 | 6 | 894 | BAB24252.2 | |
BC119174.1 | 6 | 642 | ||
BC137579.1 | 6 | 642 |