Assay Details
Target Gene Details
Entrez Gene ID: | 13350 |
Gene Name: | diacylglycerol O-acyltransferase 1 |
Gene Aliases: |
ARAT, C75990, D15Ertd23e, Dgat |
Location: |
Chr.15:76502014-76512020 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dgat1 | NM_010046.2 | NP_034176.1 | ||
AB057816.1 | BAC66171.1 | |||
AF078752.1 | AAC72917.1 | |||
BC003717.1 | AAH03717.1 |
More Information
Set Membership: |
Intragenic Non-exonic |