Assay Details
Target Gene Details
Entrez Gene ID: | 12614 |
Gene Name: | cadherin, EGF LAG seven-pass G-type receptor 1 (flamingo homolog, Drosophila) |
Gene Aliases: |
Adgrc1, Crsh, Scy, crash |
Location: |
Chr.15:85898758-86034217 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Celsr1 | NM_009886.2 | NP_034016.2 | ||
AF031572.1 | AAC68836.1 | |||
AK220170.1 | BAD90355.1 | |||
BQ180311.1 |
More Information
Set Membership: |
Intragenic Non-exonic |