Assay Details
Target Gene Details
Entrez Gene ID: | 13072 |
Gene Name: | cytochrome P450, family 11, subfamily b, polypeptide 2 |
Gene Aliases: |
Cpn2, Cyp11b, Cyp11b-2 |
Location: |
Chr.15:74851010-74856318 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cyp11b2 | NM_009991.3 | NP_034121.3 | ||
BC116908.1 | ||||
BC119321.1 |